Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDCD1
Entrez Gene ID
84955
Gene Name
NudC domain containing 1
Gene Aliases
CML66, OVA66
Chromosome Location
Chr.8: 109240919 - 109334205 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128211.1 NP_001121683.1 5 - 6 832 121
NM_032869.3 NP_116258.2 5 - 6 932 121
XM_017013910.1 XP_016869399.1 6 - 7 1108 121
XM_017013911.1 XP_016869400.1 6 - 7 878 121
GenBank mRNA AF283301.1 - 5 - 6 1053 121
AF283302.1 - 5 - 6 790 121
AF521133.1 - 5 - 6 832 121
AK027897.1 - 5 - 6 884 121
AK301278.1 - 6 - 7 948 121
AK307663.1 - 5 - 6 833 121
AK311583.1 - 6 - 7 884 121
AK315006.1 - 5 - 6 901 121
AL832317.1 - 2 - 3 240 121
BC000967.2 - 5 - 6 877 121
BC031258.1 - 6 - 7 1022 121
BC043406.1 - 5 - 6 897 121

View Full Product Details