Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPYS
Entrez Gene ID
1807
Gene Name
dihydropyrimidinase
Gene Aliases
DHP, DHPase
Chromosome Location
Chr.8: 104379424 - 104467075 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001385.2 NP_001376.1 3 - 4 731 69
XM_005250818.3 XP_005250875.1 3 - 4 756 69
XM_006716518.3 XP_006716581.1 2 - 3 597 69
XM_011516903.2 XP_011515205.1 3 - 4 757 69
XM_017013167.1 XP_016868656.1 3 - 4 756 69
GenBank mRNA BC034395.1 - 3 - 4 695 69
D78011.1 - 3 - 4 731 69

View Full Product Details