Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPYS
Entrez Gene ID
1807
Gene Name
dihydropyrimidinase
Gene Aliases
DHP, DHPase
Chromosome Location
Chr.8: 104379424 - 104467075 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001385.2 NP_001376.1 9 - 10 1700 73
XM_005250818.3 XP_005250875.1 10 - 11 1833 73
XM_006716518.3 XP_006716581.1 9 - 10 1674 73
GenBank mRNA BC034395.1 - 9 - 10 1664 73
D78011.1 - 9 - 10 1700 74

View Full Product Details