The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017708.3 | NP_060178.2 | 2 - 3 | 696 | 78 |
XM_005259019.1 | XP_005259076.1 | 3 - 4 | 2020 | 78 | |
XM_011527049.2 | XP_011525351.1 | 3 - 4 | 2005 | 78 | |
GenBank mRNA | AK000207.1 | - | 2 - 3 | 822 | 78 |
BC111970.1 | - | 2 - 3 | 696 | 78 | |
BC111972.1 | - | 2 - 3 | 696 | 78 |