Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EML4
Entrez Gene ID
27436
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
C2orf2, ELP120, EMAP-4, EMAPL4, ROPP120
Chromosome Location
Chr.2: 42169338 - 42332548 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145076.2 NP_001138548.1 9 - 10 1222 99
NM_019063.4 NP_061936.2 10 - 11 1396 99
XM_005264267.1 XP_005264324.1 11 - 12 1409 99
XM_005264268.3 XP_005264325.1 11 - 12 1409 99
XM_006711991.1 XP_006712054.1 10 - 11 1235 99
XM_006711992.2 XP_006712055.1 12 - 13 1530 99
GenBank mRNA AF177377.2 - 10 - 11 1358 99
AK021782.1 - 3 - 4 245 99
AK314702.1 - 10 - 11 1370 99
BC140845.1 - 10 - 11 1319 99
BC146799.1 - 9 - 10 1196 99
BX647693.1 - 1 - 2 248 99
BX647731.1 - 1 - 2 459 99

View Full Product Details