The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033.4 | NP_001024.1 | 11 - 12 | 1451 | 118 |
NM_001318064.1 | NP_001304993.1 | 10 - 11 | 1273 | 118 | |
NM_001318065.1 | NP_001304994.1 | 5 - 6 | 798 | 118 | |
XM_011520278.1 | XP_011518580.1 | 5 - 6 | 751 | 118 | |
GenBank mRNA | AK222786.1 | - | 11 - 12 | 1331 | 118 |
AK297988.1 | - | 11 - 12 | 1196 | 118 | |
AK299629.1 | - | 9 - 10 | 1088 | 118 | |
AK301922.1 | - | 5 - 6 | 633 | 118 | |
AK303394.1 | - | 10 - 11 | 1117 | 118 | |
AK311593.1 | - | 5 - 6 | 756 | 118 | |
BC006498.2 | - | 11 - 12 | 1282 | 118 | |
X59543.1 | - | 11 - 12 | 1310 | 118 | |
X59617.1 | - | 11 - 12 | 1304 | 118 |