The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020387.3 | NP_065120.2 | 1 - 2 | 300 | 93 |
GenBank mRNA | AF083124.1 | - | 1 - 2 | 261 | 93 |
BC009831.2 | - | 1 - 2 | 274 | 93 | |
BC033322.1 | - | 1 - 2 | 264 | 93 | |
BP263750.1 | - | 1 - 2 | 300 | 93 |