Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ASAP1-IT1
- Entrez Gene ID
-
29065
- Gene Name
- ASAP1 intronic transcript 1
- Gene Aliases
-
ASAP1-IT, ASAP1IT, ASAP1IT1, DDEF1IT1, HSPC054, NCRNA00050
- Chromosome Location
-
Chr.8: 130295355 - 130296533 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002765.2 |
|
1 - 1 |
863 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ASAP1 intronic transcript 2 |
ASAP1-IT2 |
|
-210.6kb
|
Sense |
Chr. - 130082738 - 130084768 on Build GRCh38 |
Downstream |
adenylate cyclase 8 |
ADCY8 |
AC8, ADCY3, HBAC1 |
483.8kb
|
Sense |
Chr. - 130780300 - 131041604 on Build GRCh38 |
View Full Product Details