Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS1
Entrez Gene ID
9837
Gene Name
GINS complex subunit 1
Gene Aliases
PSF1
Chromosome Location
Chr.20: 25407470 - 25448568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021067.4 NP_066545.3 3 - 4 380 67
NR_134574.1 3 - 4 380 67
XM_017028162.1 XP_016883651.1 3 - 4 337 67
GenBank mRNA BC012542.1 - 3 - 4 381 67
D80008.1 - 3 - 4 336 67

View Full Product Details