The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021067.4 | NP_066545.3 | 4 - 5 | 473 | 126 |
XM_017028162.1 | XP_016883651.1 | 4 - 5 | 430 | 126 | |
GenBank mRNA | BC012542.1 | - | 4 - 5 | 474 | 126 |
D80008.1 | - | 4 - 5 | 429 | 126 |