The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014780.4 | NP_055595.2 | 2 - 3 | 911 | 75 |
XM_005249503.2 | XP_005249560.1 | 2 - 3 | 774 | 75 | |
XM_011515020.2 | XP_011513322.1 | 2 - 3 | 772 | 75 | |
XM_017011535.1 | XP_016867024.1 | 2 - 3 | 773 | 75 | |
XM_017011538.1 | XP_016867027.1 | 2 - 3 | 916 | 75 | |
XM_017011539.1 | XP_016867028.1 | 2 - 3 | 916 | 75 | |
GenBank mRNA | AK292806.1 | - | 2 - 3 | 848 | 75 |
BC033647.1 | - | 2 - 3 | 893 | 75 | |
D38548.1 | - | 2 - 3 | 665 | 75 |