Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUL7
Entrez Gene ID
9820
Gene Name
cullin 7
Gene Aliases
3M1, KIAA0076, dJ20C7.5
Chromosome Location
Chr.6: 43037617 - 43053950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168370.1 NP_001161842.1 5 - 6 1712 84
NM_014780.4 NP_055595.2 5 - 6 1705 84
XM_005249503.2 XP_005249560.1 5 - 6 1568 84
XM_006715285.1 XP_006715348.1 5 - 6 1806 84
XM_011515019.2 XP_011513321.1 5 - 6 1663 84
XM_011515020.2 XP_011513322.1 5 - 6 1566 84
XM_017011533.1 XP_016867022.1 5 - 6 1664 84
XM_017011534.1 XP_016867023.1 5 - 6 1663 84
XM_017011535.1 XP_016867024.1 5 - 6 1567 84
XM_017011536.1 XP_016867025.1 5 - 6 1806 84
XM_017011537.1 XP_016867026.1 5 - 6 1806 84
XM_017011538.1 XP_016867027.1 5 - 6 1710 84
XM_017011539.1 XP_016867028.1 5 - 6 1710 84
XM_017011540.1 XP_016867029.1 5 - 6 1669 84
GenBank mRNA AK292806.1 - 5 - 6 1642 84
AK302668.1 - 5 - 6 1649 84
BC033647.1 - 5 - 6 1687 84
D38548.1 - 5 - 6 1459 84

View Full Product Details