The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000174.4 | NP_000165.1 | 2 - 3 | 214 | 83 |
XM_005247374.3 | XP_005247431.1 | 4 - 5 | 510 | 83 | |
XM_011512701.1 | XP_011511003.1 | 5 - 6 | 592 | 83 | |
GenBank mRNA | BC030229.1 | - | 2 - 3 | 179 | 83 |
X52997.1 | - | 2 - 3 | 214 | 83 |