The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020750.2 | NP_065801.1 | 22 - 23 | 2756 | 84 |
GenBank mRNA | AB033117.2 | - | 22 - 23 | 2695 | 84 |
AF271159.1 | - | 22 - 23 | 2545 | 84 | |
AF298880.1 | - | 22 - 23 | 2608 | 84 | |
AK001195.1 | - | 1 - 2 | 90 | 84 | |
AK022718.1 | - | 8 - 9 | 939 | 84 | |
AK024301.1 | - | 9 - 10 | 996 | 84 | |
AK056619.1 | - | 18 - 19 | 2081 | 84 | |
AK291452.1 | - | 22 - 23 | 2719 | 84 | |
AL137467.1 | - | 10 - 11 | 1199 | 84 | |
AL834207.1 | - | 22 - 23 | 2665 | 84 | |
BC000129.1 | - | 3 - 4 | 255 | 84 | |
BC008347.1 | - | 6 - 7 | 765 | 84 | |
BC009969.2 | - | 15 - 16 | 1720 | 84 | |
BC062635.1 | - | 22 - 23 | 2722 | 84 |