The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020750.2 | NP_065801.1 | 26 - 27 | 3128 | 68 |
GenBank mRNA | AB033117.2 | - | 26 - 27 | 3067 | 68 |
AF271159.1 | - | 26 - 27 | 2917 | 68 | |
AF298880.1 | - | 26 - 27 | 2980 | 68 | |
AK001195.1 | - | 5 - 6 | 461 | 68 | |
AK022718.1 | - | 12 - 13 | 1311 | 68 | |
AK023431.1 | - | 16 - 17 | 1720 | 68 | |
AK024301.1 | - | 13 - 14 | 1368 | 68 | |
AK056619.1 | - | 22 - 23 | 2453 | 68 | |
AK291452.1 | - | 26 - 27 | 3091 | 68 | |
AL137467.1 | - | 14 - 15 | 1571 | 68 | |
AL834207.1 | - | 26 - 27 | 3037 | 68 | |
BC000129.1 | - | 7 - 8 | 627 | 68 | |
BC008347.1 | - | 10 - 11 | 1137 | 68 | |
BC009969.2 | - | 19 - 20 | 2092 | 68 | |
BC062635.1 | - | 26 - 27 | 3094 | 68 |