The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000210.3 | NP_000201.2 | 22 - 23 | 3121 | 71 |
NM_001079818.2 | NP_001073286.1 | 22 - 23 | 3121 | 71 | |
NM_001316306.1 | NP_001303235.1 | 22 - 23 | 2807 | 71 | |
XM_006712510.1 | XP_006712573.1 | 22 - 23 | 3045 | 71 | |
XM_006712511.1 | XP_006712574.1 | 22 - 23 | 3045 | 71 | |
XM_017004005.1 | XP_016859494.1 | 22 - 23 | 2817 | 71 | |
XM_017004006.1 | XP_016859495.1 | 22 - 23 | 2772 | 71 | |
XM_017004007.1 | XP_016859496.1 | 22 - 23 | 2817 | 71 | |
XM_017004008.1 | XP_016859497.1 | 22 - 23 | 2772 | 71 | |
GenBank mRNA | AB208842.1 | - | 22 - 23 | 2788 | 71 |
AK294436.1 | - | 18 - 19 | 2209 | 71 | |
AK296496.1 | - | 22 - 23 | 2743 | 71 | |
BC050585.1 | - | 16 - 17 | 1727 | 71 | |
BC136455.1 | - | 22 - 23 | 2995 | 71 | |
BC136456.1 | - | 22 - 23 | 2995 | 71 | |
S66196.1 | - | 1 - 2 | 110 | 71 | |
S66213.1 | - | 1 - 2 | 110 | 71 | |
X53586.1 | - | 22 - 23 | 3033 | 71 | |
X59512.1 | - | 22 - 23 | 2869 | 71 |