Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 92246216 - 92401773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019004.1 NP_061877.1 10 - 11 1864 82
XM_005250458.3 XP_005250515.1 10 - 11 1882 82
XM_011516334.1 XP_011514636.1 9 - 10 1699 82
XM_011516335.2 XP_011514637.1 10 - 11 1446 82
XM_017012359.1 XP_016867848.1 10 - 11 1882 82
XM_017012360.1 XP_016867849.1 6 - 7 657 82
GenBank mRNA AB037807.1 - 10 - 11 1864 82
AK025475.1 - 7 - 8 961 82
BC031483.1 - 3 - 4 393 82
BC063861.1 - 10 - 11 1840 82
BC073893.1 - 4 - 5 456 82

View Full Product Details