Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 92246216 - 92401773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019004.1 NP_061877.1 7 - 8 1454 105
XM_005250458.3 XP_005250515.1 7 - 8 1472 105
XM_011516334.1 XP_011514636.1 6 - 7 1289 105
XM_011516335.2 XP_011514637.1 7 - 8 1036 105
XM_017012359.1 XP_016867848.1 7 - 8 1472 105
GenBank mRNA AB037807.1 - 7 - 8 1454 105
AK025475.1 - 4 - 5 551 105
BC063861.1 - 7 - 8 1430 105
BC073893.1 - 1 - 2 46 105

View Full Product Details