The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019004.1 | NP_061877.1 | 8 - 9 | 1609 | 84 |
XM_005250458.3 | XP_005250515.1 | 8 - 9 | 1627 | 84 | |
XM_011516334.1 | XP_011514636.1 | 7 - 8 | 1444 | 84 | |
XM_011516335.2 | XP_011514637.1 | 8 - 9 | 1191 | 84 | |
XM_017012359.1 | XP_016867848.1 | 8 - 9 | 1627 | 84 | |
XM_017012360.1 | XP_016867849.1 | 4 - 5 | 402 | 84 | |
GenBank mRNA | AB037807.1 | - | 8 - 9 | 1609 | 84 |
AK025475.1 | - | 5 - 6 | 706 | 84 | |
BC031483.1 | - | 1 - 2 | 138 | 84 | |
BC063861.1 | - | 8 - 9 | 1585 | 84 | |
BC073893.1 | - | 2 - 3 | 201 | 84 |