Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPM6B
Entrez Gene ID
2824
Gene Name
glycoprotein M6B
Gene Aliases
M6B
Chromosome Location
Chr.X: 13770922 - 13938824 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001995.2 NP_001001995.1 1 - 2 521 73
NM_001001996.2 NP_001001996.1 1 - 2 521 73
GenBank mRNA AK095657.1 - 1 - 2 275 73
BC047295.1 - 1 - 2 231 73

View Full Product Details