The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002333.3 | NP_002324.2 | 5 - 6 | 1686 | 69 |
GenBank mRNA | AB009462.1 | - | 5 - 6 | 1819 | 69 |
AK074751.1 | - | 3 - 4 | 1402 | 69 | |
AK126750.1 | - | 1 - 2 | 2494 | 69 | |
AK296536.1 | - | 5 - 6 | 1567 | 69 | |
AK316314.1 | - | 4 - 5 | 1513 | 69 | |
BC007408.2 | - | 5 - 6 | 1615 | 69 |