The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023940.2 | NP_076429.1 | 1 - 2 | 323 | 89 |
GenBank mRNA | AK314942.1 | - | 1 - 2 | 334 | 89 |
AY839725.1 | - | 1 - 2 | 334 | 89 | |
AY839726.1 | - | 1 - 2 | 334 | 89 | |
BC001087.2 | - | 1 - 2 | 323 | 89 | |
BC001846.2 | - | 1 - 2 | 323 | 89 | |
BC025694.1 | - | 1 - 2 | 322 | 89 |