The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257389.1 | NP_001244318.1 | 4 - 5 | 730 | 73 |
NM_001257390.1 | NP_001244319.1 | 4 - 5 | 473 | 73 | |
NM_001257391.1 | NP_001244320.1 | 3 - 4 | 468 | 73 | |
NM_001257392.1 | NP_001244321.1 | 3 - 4 | 269 | 73 | |
NM_001257400.1 | NP_001244329.1 | 3 - 4 | 298 | 73 | |
NM_001257401.1 | NP_001244330.1 | 3 - 4 | 438 | 73 | |
NM_001267698.1 | NP_001254627.1 | 4 - 5 | 520 | 73 | |
NM_001780.5 | NP_001771.1 | 4 - 5 | 524 | 73 | |
GenBank mRNA | AF508304.1 | - | 3 - 4 | 266 | 73 |
AK311893.1 | - | 4 - 5 | 395 | 73 | |
AL582859.3 | - | 3 - 4 | 343 | 73 | |
BC002349.2 | - | 4 - 5 | 412 | 73 | |
BC013017.2 | - | 4 - 5 | 368 | 73 | |
BE407235.1 | - | 3 - 4 | 467 | 73 | |
BG478842.1 | - | 4 - 5 | 474 | 73 | |
BQ434569.1 | - | - | 439 | 73 | |
BT007073.1 | - | 3 - 4 | 327 | 73 | |
BT020137.1 | - | 3 - 4 | 327 | 73 | |
BT020138.1 | - | 3 - 4 | 327 | 73 | |
BU167765.1 | - | 4 - 5 | 531 | 73 | |
BU190521.1 | - | 3 - 4 | 463 | 73 | |
CN296813.1 | - | 4 - 5 | 487 | 73 | |
CN484590.1 | - | 3 - 4 | 338 | 73 | |
CR542096.1 | - | 3 - 4 | 327 | 73 | |
EB386351.1 | - | 4 - 5 | 459 | 73 | |
KF998086.1 | - | 3 - 4 | 327 | 73 | |
M58485.1 | - | 4 - 5 | 382 | 73 | |
M59907.1 | - | 4 - 5 | 377 | 73 | |
S93788.1 | - | 3 - 4 | 351 | 73 | |
X07982.1 | - | 4 - 5 | 354 | 73 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BC071882.1 | - | 949 | Chr. - - [-] on Build | 73 |