The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006832.2 | NP_006823.1 | 12 - 13 | 1786 | 74 |
XM_005267285.2 | XP_005267342.1 | 13 - 14 | 1914 | 74 | |
GenBank mRNA | AF443279.1 | - | 12 - 13 | 1681 | 74 |
AK291738.1 | - | 12 - 13 | 1739 | 74 | |
BC017327.2 | - | 12 - 13 | 1786 | 74 | |
Z24725.1 | - | 12 - 13 | 1722 | 74 |