Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
Chromosome Location
Chr.14: 52857271 - 52951192 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134999.1 NP_001128471.1 7 - 8 1149 137
NM_001135000.1 NP_001128472.1 7 - 8 1149 137
NM_006832.2 NP_006823.1 7 - 8 1149 137
XM_005267285.2 XP_005267342.1 8 - 9 1277 137
XM_006720008.2 XP_006720071.1 8 - 9 1277 137
XM_006720009.2 XP_006720072.1 8 - 9 1277 137
XM_006720010.2 XP_006720073.1 7 - 8 1244 137
GenBank mRNA AF443279.1 - 7 - 8 1044 137
AK291738.1 - 7 - 8 1102 137
BC017327.2 - 7 - 8 1149 137
BX161467.1 - 7 - 8 1070 137
EU979385.1 - 6 - 7 963 137
Z24725.1 - 7 - 8 1085 137

View Full Product Details