The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320833.1 | NP_001307762.1 | 3 - 4 | 622 | 120 |
NM_001320834.1 | NP_001307763.1 | 2 - 3 | 191 | 120 | |
NM_152386.3 | NP_689599.2 | 2 - 3 | 479 | 120 | |
XM_017003355.1 | XP_016858844.1 | 2 - 3 | 381 | 120 | |
GenBank mRNA | AF542512.1 | - | 2 - 3 | 383 | 120 |
AK096323.1 | - | 2 - 3 | 193 | 120 | |
AK314322.1 | - | 2 - 3 | 479 | 120 | |
BC106904.1 | - | 2 - 3 | 374 | 120 | |
BC106905.1 | - | 2 - 3 | 376 | 120 | |
BC134342.1 | - | 2 - 3 | 383 | 120 |