The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144999.2 | NP_001138471.1 | 15 - 16 | 1492 | 73 |
NM_001145000.2 | NP_001138472.1 | 13 - 14 | 1668 | 73 | |
NM_002210.4 | NP_002201.1 | 15 - 16 | 1776 | 73 | |
GenBank mRNA | AB209894.1 | - | 13 - 14 | 1651 | 73 |
AK302990.1 | - | 15 - 16 | 1492 | 73 | |
BC126231.1 | - | 15 - 16 | 1629 | 73 | |
BC136442.1 | - | 15 - 16 | 1629 | 73 | |
BC144100.1 | - | 13 - 14 | 1521 | 73 | |
EF560727.1 | - | 15 - 16 | 1544 | 73 | |
M14648.1 | - | 15 - 16 | 1541 | 73 |