The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242463.1 | NP_001229392.1 | 5 - 6 | 795 | 70 |
NM_058229.3 | NP_478136.1 | 7 - 8 | 1074 | 70 | |
NM_148177.2 | NP_680482.1 | 4 - 5 | 709 | 70 | |
GenBank mRNA | AJ420108.1 | - | 7 - 8 | 924 | 70 |
AK056986.1 | - | 7 - 8 | 1027 | 70 | |
AY059629.1 | - | 7 - 8 | 835 | 70 | |
BC024030.2 | - | 4 - 5 | 709 | 70 | |
BC100020.1 | - | 6 - 7 | 665 | 70 | |
BC120963.1 | - | 4 - 5 | 703 | 70 | |
BC120964.1 | - | 4 - 5 | 703 | 70 | |
EF143260.1 | - | 5 - 6 | 748 | 70 |