The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290046.1 | NP_001276975.1 | 11 - 12 | 1229 | 125 |
NM_001290047.1 | NP_001276976.1 | 11 - 12 | 1698 | 125 | |
XM_011546128.2 | XP_011544430.1 | 10 - 11 | 1302 | 125 | |
XM_011546129.2 | XP_011544431.1 | 11 - 12 | 1386 | 125 | |
XM_011546132.2 | XP_011544434.1 | 11 - 12 | 1386 | 125 | |
XM_017028785.1 | XP_016884274.1 | 11 - 12 | 1386 | 125 | |
GenBank mRNA | AB051527.1 | - | 3 - 4 | 247 | 125 |
AF336133.1 | - | 11 - 12 | 1758 | 125 | |
AF411609.1 | - | 7 - 8 | 755 | 125 | |
AK308330.1 | - | 11 - 12 | 1230 | 125 |