Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRISP2
Entrez Gene ID
7180
Gene Name
cysteine rich secretory protein 2
Gene Aliases
CRISP-2, CT36, GAPDL5, TPX1, TSP1
Chromosome Location
Chr.6: 49676665 - 49714198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142407.2 NP_001135879.1 6 - 7 618 114
NM_001142408.2 NP_001135880.1 7 - 8 675 114
NM_001142417.2 NP_001135889.1 7 - 8 655 114
NM_001142435.2 NP_001135907.1 6 - 7 638 114
NM_001261822.1 NP_001248751.1 6 - 7 644 114
NM_003296.3 NP_003287.1 7 - 8 681 114
XM_005249349.1 XP_005249406.1 7 - 8 655 114
XM_005249350.1 XP_005249407.1 6 - 7 613 114
XM_005249351.3 XP_005249408.1 6 - 7 633 114
XM_005249352.3 XP_005249409.1 6 - 7 624 114
XM_005249353.3 XP_005249410.1 7 - 8 660 114
XM_005249356.1 XP_005249413.1 7 - 8 658 114
XM_011514841.1 XP_011513143.1 7 - 8 990 114
XM_011514842.1 XP_011513144.1 6 - 7 953 114
XM_011514843.2 XP_011513145.1 7 - 8 660 114
GenBank mRNA AK223337.1 - 7 - 8 618 114
AK292385.1 - 7 - 8 659 114
AY289796.1 - 7 - 8 625 114
AY292862.1 - 6 - 7 615 114
AY292863.1 - 6 - 7 619 114
AY773479.1 - 7 - 8 661 114
BC022011.2 - 7 - 8 628 114
BC107707.1 - 6 - 7 575 114
BG718726.1 - 6 - 7 614 114
BM552843.1 - 6 - 7 553 114
BP369727.1 - 1 - 2 132 114
M25532.1 - 7 - 8 604 114
X95239.1 - 7 - 8 655 114

View Full Product Details