Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFDP1
Entrez Gene ID
10428
Gene Name
craniofacial development protein 1
Gene Aliases
BCNT, BUCENTAUR, CENP-29, CP27, SWC5, Yeti, p97
Chromosome Location
Chr.16: 75293710 - 75433512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006324.2 NP_006315.1 4 - 5 666 133
XM_011522814.1 XP_011521116.1 4 - 5 689 133
XM_011522815.1 XP_011521117.1 4 - 5 689 133
GenBank mRNA AB009285.1 - 4 - 5 638 133
AK226031.1 - 5 - 6 654 133
BC000991.2 - 4 - 5 662 133
BT009819.1 - 4 - 5 529 133
CR542111.1 - 4 - 5 529 133
D85939.1 - 4 - 5 638 133
D86549.1 - 3 - 4 465 133
LC057679.1 - 4 - 5 529 133
LC057680.1 - 3 - 4 411 133
LC057682.1 - 4 - 5 529 133
LC057683.1 - 4 - 5 529 133
LC057684.1 - 3 - 4 411 133

View Full Product Details