The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001254718.1 | NP_001241647.1 | 10 - 11 | 1025 | 101 |
NM_001254719.1 | NP_001241648.1 | 10 - 11 | 1025 | 101 | |
NM_001254720.1 | NP_001241649.1 | 9 - 10 | 989 | 101 | |
NM_001254721.1 | NP_001241650.1 | 9 - 10 | 968 | 101 | |
NM_001254722.1 | NP_001241651.1 | 8 - 9 | 947 | 101 | |
NM_001254723.1 | NP_001241652.1 | 9 - 10 | 986 | 101 | |
NM_002465.3 | NP_002456.2 | 12 - 13 | 1100 | 101 | |
NM_206819.2 | NP_996555.1 | 12 - 13 | 1100 | 101 | |
NM_206820.2 | NP_996556.1 | 10 - 11 | 1025 | 101 | |
NM_206821.2 | NP_996557.1 | 10 - 11 | 1025 | 101 | |
XM_005268876.4 | XP_005268933.1 | 11 - 12 | 1034 | 101 | |
XM_006719405.3 | XP_006719468.1 | 12 - 13 | 961 | 101 | |
XM_006719406.3 | XP_006719469.1 | 12 - 13 | 961 | 101 | |
XM_006719407.3 | XP_006719470.1 | 11 - 12 | 1034 | 101 | |
XM_006719408.3 | XP_006719471.1 | 12 - 13 | 961 | 101 | |
XM_006719409.3 | XP_006719472.1 | 10 - 11 | 987 | 101 | |
XM_006719410.3 | XP_006719473.1 | 12 - 13 | 961 | 101 | |
XM_006719411.3 | XP_006719474.1 | 10 - 11 | 987 | 101 | |
XM_017019315.1 | XP_016874804.1 | 11 - 12 | 904 | 101 | |
XM_017019316.1 | XP_016874805.1 | 12 - 13 | 961 | 101 | |
XM_017019317.1 | XP_016874806.1 | 10 - 11 | 987 | 101 | |
XM_017019318.1 | XP_016874807.1 | 11 - 12 | 904 | 101 | |
XM_017019319.1 | XP_016874808.1 | 9 - 10 | 930 | 101 | |
XM_017019320.1 | XP_016874809.1 | 8 - 9 | 1029 | 101 | |
XM_017019321.1 | XP_016874810.1 | 9 - 10 | 930 | 101 | |
XM_017019322.1 | XP_016874811.1 | 7 - 8 | 972 | 101 | |
GenBank mRNA | AK056587.1 | - | 11 - 12 | 1024 | 101 |
AK292976.1 | - | 12 - 13 | 1060 | 101 | |
AK295727.1 | - | 8 - 9 | 907 | 101 | |
AK296681.1 | - | 10 - 11 | 989 | 101 | |
AK303401.1 | - | 9 - 10 | 946 | 101 | |
AL831993.2 | - | 12 - 13 | 1035 | 101 | |
AL832000.2 | - | 12 - 13 | 1035 | 101 | |
AL832332.2 | - | 12 - 13 | 1035 | 101 | |
AL832350.2 | - | 12 - 13 | 1035 | 101 | |
AL832456.2 | - | 12 - 13 | 1035 | 101 | |
AL834249.2 | - | 12 - 13 | 1035 | 101 | |
BC015187.1 | - | 12 - 13 | 1060 | 101 | |
BC065720.1 | - | 8 - 9 | 910 | 101 | |
BC092418.1 | - | 10 - 11 | 972 | 101 | |
BC117217.1 | - | 8 - 9 | 882 | 101 | |
BC143495.1 | - | 9 - 10 | 903 | 101 | |
BC143502.1 | - | 8 - 9 | 882 | 101 | |
BC143503.1 | - | 10 - 11 | 960 | 101 | |
BC143504.1 | - | 9 - 10 | 924 | 101 | |
BX649073.2 | - | 13 - 14 | 1131 | 101 | |
X73114.1 | - | 10 - 11 | 966 | 101 |