Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
Chromosome Location
Chr.12: 101594849 - 101695841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254718.1 NP_001241647.1 13 - 14 1424 110
NM_001254719.1 NP_001241648.1 13 - 14 1424 110
NM_001254720.1 NP_001241649.1 12 - 13 1388 110
NM_001254721.1 NP_001241650.1 12 - 13 1367 110
NM_001254722.1 NP_001241651.1 11 - 12 1346 110
NM_001254723.1 NP_001241652.1 12 - 13 1385 110
NM_002465.3 NP_002456.2 15 - 16 1499 110
NM_206819.2 NP_996555.1 15 - 16 1499 110
NM_206820.2 NP_996556.1 13 - 14 1424 110
NM_206821.2 NP_996557.1 13 - 14 1424 110
XM_005268876.4 XP_005268933.1 14 - 15 1433 110
XM_006719405.3 XP_006719468.1 15 - 16 1360 110
XM_006719406.3 XP_006719469.1 15 - 16 1360 110
XM_006719407.3 XP_006719470.1 14 - 15 1433 110
XM_006719408.3 XP_006719471.1 15 - 16 1360 110
XM_006719409.3 XP_006719472.1 13 - 14 1386 110
XM_006719410.3 XP_006719473.1 15 - 16 1360 110
XM_006719411.3 XP_006719474.1 13 - 14 1386 110
XM_017019315.1 XP_016874804.1 14 - 15 1303 110
XM_017019316.1 XP_016874805.1 15 - 16 1360 110
XM_017019317.1 XP_016874806.1 13 - 14 1386 110
XM_017019318.1 XP_016874807.1 14 - 15 1303 110
XM_017019319.1 XP_016874808.1 12 - 13 1329 110
XM_017019320.1 XP_016874809.1 11 - 12 1428 110
XM_017019321.1 XP_016874810.1 12 - 13 1329 110
XM_017019322.1 XP_016874811.1 10 - 11 1371 110
GenBank mRNA AK292976.1 - 15 - 16 1459 110
AK295727.1 - 11 - 12 1306 110
AK296681.1 - 13 - 14 1388 110
AK303401.1 - 12 - 13 1345 110
AL831993.2 - 15 - 16 1434 110
AL832332.2 - 15 - 16 1434 110
AL832350.2 - 15 - 16 1434 110
AL832354.2 - 15 - 16 1436 110
AL834249.2 - 15 - 16 1434 110
BC015187.1 - 15 - 16 1459 110
BC065720.1 - 11 - 12 1309 110
BC092418.1 - 13 - 14 1371 110
BC117217.1 - 11 - 12 1281 110
BC143495.1 - 12 - 13 1302 110
BC143502.1 - 11 - 12 1281 110
BC143503.1 - 13 - 14 1359 110
BC143504.1 - 12 - 13 1323 110
BX649073.2 - 16 - 17 1530 110
X66276.1 - 13 - 14 1374 110

View Full Product Details