Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
Chromosome Location
Chr.12: 101594849 - 101695841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254718.1 NP_001241647.1 14 - 15 1591 66
NM_001254719.1 NP_001241648.1 14 - 15 1591 66
NM_001254720.1 NP_001241649.1 13 - 14 1555 66
NM_001254721.1 NP_001241650.1 13 - 14 1534 66
NM_001254722.1 NP_001241651.1 12 - 13 1513 66
NM_001254723.1 NP_001241652.1 13 - 14 1552 66
NM_002465.3 NP_002456.2 16 - 17 1666 66
NM_206819.2 NP_996555.1 16 - 17 1666 66
NM_206820.2 NP_996556.1 14 - 15 1591 66
NM_206821.2 NP_996557.1 14 - 15 1591 66
XM_005268876.4 XP_005268933.1 15 - 16 1600 66
XM_006719405.3 XP_006719468.1 16 - 17 1527 66
XM_006719406.3 XP_006719469.1 16 - 17 1527 66
XM_006719407.3 XP_006719470.1 15 - 16 1600 66
XM_006719408.3 XP_006719471.1 16 - 17 1527 66
XM_006719409.3 XP_006719472.1 14 - 15 1553 66
XM_006719410.3 XP_006719473.1 16 - 17 1527 66
XM_006719411.3 XP_006719474.1 14 - 15 1553 66
XM_017019315.1 XP_016874804.1 15 - 16 1470 66
XM_017019316.1 XP_016874805.1 16 - 17 1527 66
XM_017019317.1 XP_016874806.1 14 - 15 1553 66
XM_017019318.1 XP_016874807.1 15 - 16 1470 66
XM_017019319.1 XP_016874808.1 13 - 14 1496 66
XM_017019320.1 XP_016874809.1 12 - 13 1595 66
XM_017019321.1 XP_016874810.1 13 - 14 1496 66
XM_017019322.1 XP_016874811.1 11 - 12 1538 66
GenBank mRNA AK292976.1 - 16 - 17 1626 66
AK295727.1 - 12 - 13 1473 66
AK296681.1 - 14 - 15 1555 66
AK303401.1 - 13 - 14 1512 66
AL831993.2 - 16 - 17 1601 66
AL832000.2 - 16 - 17 1601 66
AL832332.2 - 16 - 17 1601 66
AL832354.2 - 16 - 17 1603 66
AL832456.2 - 16 - 17 1601 66
AL834249.2 - 16 - 17 1601 66
BC015187.1 - 16 - 17 1626 66
BC092418.1 - 14 - 15 1538 66
BC117217.1 - 12 - 13 1448 66
BC143495.1 - 13 - 14 1469 66
BC143502.1 - 12 - 13 1448 66
BC143503.1 - 14 - 15 1526 66
BC143504.1 - 13 - 14 1490 66
BX649073.2 - 17 - 18 1697 66
X73114.1 - 14 - 15 1532 66

View Full Product Details