Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
Chromosome Location
Chr.12: 101594849 - 101695841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254718.1 NP_001241647.1 4 - 5 345 98
NM_001254719.1 NP_001241648.1 4 - 5 345 98
NM_001254720.1 NP_001241649.1 3 - 4 309 98
NM_001254721.1 NP_001241650.1 4 - 5 345 98
NM_001254722.1 NP_001241651.1 2 - 3 267 98
NM_001254723.1 NP_001241652.1 3 - 4 306 98
NM_002465.3 NP_002456.2 6 - 7 420 98
NM_206819.2 NP_996555.1 6 - 7 420 98
NM_206820.2 NP_996556.1 4 - 5 345 98
NM_206821.2 NP_996557.1 4 - 5 345 98
XM_005268876.4 XP_005268933.1 5 - 6 354 98
XM_006719405.3 XP_006719468.1 6 - 7 281 98
XM_006719406.3 XP_006719469.1 6 - 7 281 98
XM_006719407.3 XP_006719470.1 5 - 6 354 98
XM_006719408.3 XP_006719471.1 6 - 7 281 98
XM_006719409.3 XP_006719472.1 4 - 5 307 98
XM_006719410.3 XP_006719473.1 6 - 7 281 98
XM_006719411.3 XP_006719474.1 4 - 5 307 98
XM_017019315.1 XP_016874804.1 6 - 7 281 98
XM_017019316.1 XP_016874805.1 6 - 7 281 98
XM_017019317.1 XP_016874806.1 4 - 5 307 98
XM_017019318.1 XP_016874807.1 6 - 7 281 98
XM_017019319.1 XP_016874808.1 4 - 5 307 98
XM_017019320.1 XP_016874809.1 2 - 3 349 98
XM_017019321.1 XP_016874810.1 4 - 5 307 98
XM_017019322.1 XP_016874811.1 2 - 3 349 98
GenBank mRNA AK056587.1 - 5 - 6 344 98
AK292976.1 - 6 - 7 380 98
AK295727.1 - 2 - 3 227 98
AK296681.1 - 4 - 5 309 98
AL831993.2 - 6 - 7 355 98
AL832000.2 - 6 - 7 355 98
AL832332.2 - 6 - 7 355 98
AL832354.2 - 6 - 7 357 98
AL832456.2 - 6 - 7 355 98
AL834249.2 - 6 - 7 355 98
BC015187.1 - 6 - 7 380 98
BC065720.1 - 2 - 3 230 98
BC092418.1 - 4 - 5 292 98
BC117217.1 - 2 - 3 202 98
BC143495.1 - 4 - 5 280 98
BC143502.1 - 2 - 3 202 98
BC143503.1 - 4 - 5 280 98
BC143504.1 - 3 - 4 244 98
BP232990.1 - 4 - 5 305 98
BX649073.2 - 7 - 8 451 98
DA433791.1 - 4 - 5 309 98
X73114.1 - 4 - 5 286 98

View Full Product Details