The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020715.2 | NP_065766.1 | 20 - 21 | 2989 | 71 |
XM_011537009.1 | XP_011535311.1 | 20 - 21 | 3165 | 71 | |
XM_011537015.2 | XP_011535317.1 | 13 - 14 | 3220 | 71 | |
XM_017021499.1 | XP_016876988.1 | 19 - 20 | 3146 | 71 | |
XM_017021500.1 | XP_016876989.1 | 20 - 21 | 3165 | 71 | |
XM_017021501.1 | XP_016876990.1 | 19 - 20 | 3062 | 71 | |
XM_017021502.1 | XP_016876991.1 | 13 - 14 | 2942 | 71 | |
XM_017021503.1 | XP_016876992.1 | 13 - 14 | 2302 | 71 | |
XM_017021504.1 | XP_016876993.1 | 15 - 16 | 2444 | 71 | |
GenBank mRNA | AB033026.1 | - | 20 - 21 | 2965 | 71 |
AK293950.1 | - | 1 - 2 | 67 | 71 | |
AK295896.1 | - | 13 - 14 | 1689 | 71 | |
BC146787.1 | - | 20 - 21 | 2965 | 71 | |
BX648464.1 | - | 11 - 12 | 2101 | 71 |