The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020715.2 | NP_065766.1 | 23 - 24 | 3433 | 61 |
XM_011537009.1 | XP_011535311.1 | 23 - 24 | 3609 | 61 | |
XM_011537015.2 | XP_011535317.1 | 16 - 17 | 3664 | 61 | |
XM_017021499.1 | XP_016876988.1 | 22 - 23 | 3590 | 61 | |
XM_017021500.1 | XP_016876989.1 | 23 - 24 | 3525 | 61 | |
XM_017021501.1 | XP_016876990.1 | 22 - 23 | 3506 | 61 | |
XM_017021502.1 | XP_016876991.1 | 16 - 17 | 3386 | 61 | |
XM_017021503.1 | XP_016876992.1 | 16 - 17 | 2746 | 61 | |
XM_017021504.1 | XP_016876993.1 | 18 - 19 | 2888 | 61 | |
GenBank mRNA | AB033026.1 | - | 23 - 24 | 3409 | 61 |
AK295896.1 | - | 16 - 17 | 2133 | 61 | |
BC146787.1 | - | 23 - 24 | 3409 | 61 | |
BX648464.1 | - | 14 - 15 | 2545 | 61 |