The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258206.1 | NP_001245135.1 | 9 - 10 | 1064 | 56 |
NM_001258207.1 | NP_001245136.1 | 8 - 9 | 968 | 56 | |
NM_005428.3 | NP_005419.2 | 9 - 10 | 1064 | 56 | |
XM_005259642.1 | XP_005259699.1 | 9 - 10 | 1024 | 56 | |
GenBank mRNA | AK225710.1 | - | 9 - 10 | 1018 | 56 |
AK301128.1 | - | 8 - 9 | 678 | 56 | |
AK313307.1 | - | 9 - 10 | 1006 | 56 | |
BC013361.2 | - | 9 - 10 | 844 | 56 | |
X16316.1 | - | 8 - 9 | 890 | 56 | |
X83931.1 | - | 1 - 2 | 30 | 56 |