The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006037.3 | NP_006028.2 | 15 - 16 | 2890 | 111 |
XM_006712877.3 | XP_006712940.1 | 14 - 15 | 8836 | 111 | |
XM_006712878.3 | XP_006712941.1 | 15 - 16 | 2410 | 111 | |
XM_006712879.3 | XP_006712942.1 | 14 - 15 | 8303 | 111 | |
XM_006712880.3 | XP_006712943.1 | 14 - 15 | 8315 | 111 | |
XM_011512217.2 | XP_011510519.1 | 14 - 15 | 8851 | 111 | |
XM_011512218.2 | XP_011510520.1 | 14 - 15 | 8851 | 111 | |
XM_011512219.2 | XP_011510521.1 | 14 - 15 | 8836 | 111 | |
XM_011512220.2 | XP_011510522.1 | 14 - 15 | 2590 | 111 | |
XM_011512221.1 | XP_011510523.1 | 15 - 16 | 2578 | 111 | |
XM_011512222.2 | XP_011510524.1 | 15 - 16 | 2879 | 111 | |
XM_011512223.2 | XP_011510525.1 | 15 - 16 | 2393 | 111 | |
XM_011512224.2 | XP_011510526.1 | 14 - 15 | 2781 | 111 | |
XM_011512225.2 | XP_011510527.1 | 14 - 15 | 2766 | 111 | |
XM_011512226.2 | XP_011510528.1 | 14 - 15 | 2338 | 111 | |
XM_011512227.2 | XP_011510529.1 | 13 - 14 | 7865 | 111 | |
XM_011512230.1 | XP_011510532.1 | 9 - 10 | 1776 | 111 | |
XM_017005394.1 | XP_016860883.1 | 13 - 14 | 8720 | 111 | |
XM_017005395.1 | XP_016860884.1 | 11 - 12 | 1642 | 111 | |
GenBank mRNA | AB006626.2 | - | 15 - 16 | 2890 | 111 |
AF132607.1 | - | 14 - 15 | 2098 | 111 | |
AK302719.1 | - | 9 - 10 | 1459 | 111 | |
AK303368.1 | - | 14 - 15 | 2457 | 111 | |
AK303467.1 | - | 13 - 14 | 2097 | 111 | |
AK303617.1 | - | 12 - 13 | 2071 | 111 | |
AK308791.1 | - | 15 - 16 | 2583 | 111 | |
AK310603.1 | - | 14 - 15 | 2550 | 111 | |
BC039904.1 | - | 15 - 16 | 2157 | 111 |