The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006037.3 | NP_006028.2 | 3 - 4 | 888 | 93 |
XM_006712877.3 | XP_006712940.1 | 2 - 3 | 6834 | 93 | |
XM_006712878.3 | XP_006712941.1 | 3 - 4 | 393 | 93 | |
XM_006712879.3 | XP_006712942.1 | 2 - 3 | 6286 | 93 | |
XM_006712880.3 | XP_006712943.1 | 2 - 3 | 6298 | 93 | |
XM_011512217.2 | XP_011510519.1 | 2 - 3 | 6834 | 93 | |
XM_011512218.2 | XP_011510520.1 | 2 - 3 | 6834 | 93 | |
XM_011512219.2 | XP_011510521.1 | 2 - 3 | 6834 | 93 | |
XM_011512220.2 | XP_011510522.1 | 2 - 3 | 573 | 93 | |
XM_011512221.1 | XP_011510523.1 | 3 - 4 | 561 | 93 | |
XM_011512222.2 | XP_011510524.1 | 3 - 4 | 862 | 93 | |
XM_011512223.2 | XP_011510525.1 | 3 - 4 | 376 | 93 | |
XM_011512224.2 | XP_011510526.1 | 2 - 3 | 764 | 93 | |
XM_011512225.2 | XP_011510527.1 | 2 - 3 | 764 | 93 | |
XM_017005394.1 | XP_016860883.1 | 2 - 3 | 6835 | 93 | |
GenBank mRNA | AB006626.2 | - | 3 - 4 | 888 | 93 |
AF132607.1 | - | 2 - 3 | 96 | 93 | |
AK303368.1 | - | 2 - 3 | 440 | 93 | |
AK308791.1 | - | 3 - 4 | 566 | 93 | |
AK310603.1 | - | 2 - 3 | 548 | 93 |