The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018266.2 | NP_060736.1 | 3 - 4 | 750 | 132 |
NR_073506.1 | 3 - 4 | 750 | 132 | ||
XM_005247578.1 | XP_005247635.1 | 3 - 4 | 715 | 132 | |
XM_006713687.1 | XP_006713750.1 | 3 - 4 | 658 | 132 | |
XM_011512958.2 | XP_011511260.1 | 3 - 4 | 715 | 132 | |
GenBank mRNA | AK001764.1 | - | 3 - 4 | 692 | 132 |
AK222950.1 | - | 3 - 4 | 682 | 132 | |
AK223248.1 | - | 3 - 4 | 682 | 132 | |
BC003192.1 | - | 3 - 4 | 665 | 132 | |
BC010070.2 | - | 3 - 4 | 665 | 132 | |
BC021277.2 | - | 3 - 4 | 668 | 132 |