Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIWIL1
Entrez Gene ID
9271
Gene Name
piwi like RNA-mediated gene silencing 1
Gene Aliases
CT80.1, HIWI, MIWI, PIWI
Chromosome Location
Chr.12: 130337887 - 130412708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190971.1 NP_001177900.1 13 - 14 1867 90
NM_004764.4 NP_004755.2 13 - 14 1867 90
XM_011539002.2 XP_011537304.1 13 - 14 1741 90
XM_011539003.2 XP_011537305.1 13 - 14 1672 90
XM_011539004.2 XP_011537306.1 13 - 14 1810 90
XM_011539005.1 XP_011537307.1 13 - 14 1775 90
XM_011539006.2 XP_011537308.1 5 - 6 667 90
XM_017020229.1 XP_016875718.1 13 - 14 1775 90
GenBank mRNA AF104260.2 - 13 - 14 1652 90
AF264004.1 - 10 - 11 1338 90
AF387507.1 - 13 - 14 1659 90
AK093133.1 - 13 - 14 1649 90
BC028581.2 - 13 - 14 1664 90

View Full Product Details