The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004764.4 | NP_004755.2 | 20 - 21 | 2744 | 64 |
XM_011539002.2 | XP_011537304.1 | 20 - 21 | 2618 | 64 | |
XM_011539003.2 | XP_011537305.1 | 20 - 21 | 2549 | 64 | |
XM_011539004.2 | XP_011537306.1 | 20 - 21 | 2687 | 64 | |
XM_011539005.1 | XP_011537307.1 | 19 - 20 | 2526 | 64 | |
XM_011539006.2 | XP_011537308.1 | 12 - 13 | 1544 | 64 | |
GenBank mRNA | AF104260.2 | - | 20 - 21 | 2529 | 64 |
AF264004.1 | - | 17 - 18 | 2215 | 64 | |
AF387507.1 | - | 20 - 21 | 2536 | 64 | |
BC028581.2 | - | 20 - 21 | 2541 | 64 |