Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIWIL1
Entrez Gene ID
9271
Gene Name
piwi like RNA-mediated gene silencing 1
Gene Aliases
CT80.1, HIWI, MIWI, PIWI
Chromosome Location
Chr.12: 130337887 - 130412708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190971.1 NP_001177900.1 4 - 5 586 82
NM_004764.4 NP_004755.2 4 - 5 586 82
XM_011539002.2 XP_011537304.1 4 - 5 460 82
XM_011539003.2 XP_011537305.1 4 - 5 391 82
XM_011539004.2 XP_011537306.1 4 - 5 529 82
XM_011539005.1 XP_011537307.1 4 - 5 494 82
XM_017020229.1 XP_016875718.1 4 - 5 494 82
GenBank mRNA AB274731.1 - 3 - 4 315 82
AF104260.2 - 4 - 5 371 82
AF387507.1 - 4 - 5 378 82
AK093133.1 - 4 - 5 368 82
BC028581.2 - 4 - 5 383 82

View Full Product Details