Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHP4
Entrez Gene ID
261734
Gene Name
nephronophthisis 4
Gene Aliases
POC10, SLSN4
Chromosome Location
Chr.1: 5862808 - 5992473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291593.1 NP_001278522.1 19 - 20 3192 80
NM_001291594.1 NP_001278523.1 18 - 19 3015 80
NM_015102.4 NP_055917.1 22 - 23 3502 80
NR_111987.1 25 - 26 4049 80
XM_006710563.3 XP_006710626.1 22 - 23 3348 80
XM_011541213.1 XP_011539515.1 21 - 22 3269 80
XM_011541214.1 XP_011539516.1 21 - 22 3230 80
XM_011541215.1 XP_011539517.1 20 - 21 3161 80
XM_011541216.2 XP_011539518.1 23 - 24 3976 80
XM_011541217.2 XP_011539519.1 22 - 23 3840 80
XM_011541218.2 XP_011539520.1 23 - 24 3509 80
XM_017000996.1 XP_016856485.1 21 - 22 3227 80
XM_017000997.1 XP_016856486.1 22 - 23 3771 80
XM_017000998.1 XP_016856487.1 21 - 22 3272 80
XM_017000999.1 XP_016856488.1 22 - 23 3267 80
XM_017001000.1 XP_016856489.1 21 - 22 3100 80
XM_017001001.1 XP_016856490.1 16 - 17 2539 80
XM_017001003.1 XP_016856492.1 14 - 15 2330 80
GenBank mRNA AB014573.1 - 17 - 18 2602 80
AF537130.1 - 22 - 23 3500 80
AK314326.1 - 19 - 20 3166 80
AY118228.1 - 22 - 23 3437 80
BC040520.1 - 25 - 26 4050 80

View Full Product Details