Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NPHP4
- Entrez Gene ID
-
261734
- Gene Name
- nephronophthisis 4
- Gene Aliases
-
POC10, SLSN4
- Chromosome Location
-
Chr.1: 5862808 - 5992473 on Build GRCh38
View Full Product Details