The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017572.3 | NP_060042.2 | 7 - 8 | 743 | 65 |
NM_199054.2 | NP_951009.1 | 7 - 8 | 743 | 65 | |
GenBank mRNA | AF125532.1 | - | 6 - 7 | 520 | 65 |
AF237775.1 | - | 6 - 7 | 534 | 65 | |
AF237776.1 | - | 6 - 7 | 534 | 65 | |
AK293742.1 | - | 5 - 6 | 402 | 65 | |
AL137565.1 | - | 3 - 4 | 205 | 65 | |
BC073140.1 | - | 7 - 8 | 723 | 65 |