The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018218.2 | NP_060688.1 | 18 - 19 | 2467 | 97 |
XM_006712612.3 | XP_006712675.1 | 18 - 19 | 2485 | 97 | |
XM_011511396.2 | XP_011509698.1 | 18 - 19 | 2485 | 97 | |
XM_011511397.2 | XP_011509699.1 | 19 - 20 | 2559 | 97 | |
XM_011511398.2 | XP_011509700.1 | 18 - 19 | 2485 | 97 | |
XM_011511399.2 | XP_011509701.1 | 17 - 18 | 2286 | 97 | |
XM_011511400.2 | XP_011509702.1 | 17 - 18 | 2270 | 97 | |
XM_017004426.1 | XP_016859915.1 | 19 - 20 | 2559 | 97 | |
XM_017004427.1 | XP_016859916.1 | 12 - 13 | 1487 | 97 | |
XM_017004428.1 | XP_016859917.1 | 18 - 19 | 2527 | 97 | |
GenBank mRNA | AJ583821.2 | - | 18 - 19 | 2467 | 97 |
AK001647.1 | - | 18 - 19 | 2429 | 97 | |
AK297821.1 | - | 12 - 13 | 1555 | 97 |