The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003183.5 | NP_003174.3 | 3 - 4 | 578 | 111 |
XM_011510375.2 | XP_011508677.1 | 3 - 4 | 597 | 111 | |
XM_011510376.2 | XP_011508678.1 | 3 - 4 | 546 | 111 | |
XM_017004785.1 | XP_016860274.1 | 3 - 4 | 594 | 111 | |
XM_017004786.1 | XP_016860275.1 | 2 - 3 | 266 | 111 | |
GenBank mRNA | AK289829.1 | - | 3 - 4 | 513 | 111 |
BC062687.1 | - | 3 - 4 | 532 | 111 | |
BC136783.1 | - | 3 - 4 | 440 | 111 | |
BC146658.1 | - | 3 - 4 | 550 | 111 | |
CN258241.1 | - | 3 - 4 | 548 | 111 | |
GQ402545.1 | - | 3 - 4 | 546 | 111 | |
U69611.1 | - | 3 - 4 | 477 | 111 | |
U69612.1 | - | 3 - 4 | 485 | 111 | |
U86755.1 | - | 3 - 4 | 526 | 111 |