The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015888.4 | NP_056972.1 | 19 - 20 | 2093 | 137 |
XM_011541562.2 | XP_011539864.1 | 19 - 20 | 2063 | 137 | |
GenBank mRNA | AF044923.1 | - | 19 - 20 | 2035 | 137 |
AK097735.1 | - | 17 - 18 | 1836 | 137 | |
AK098561.1 | - | 13 - 14 | 1374 | 137 | |
AK292314.1 | - | 20 - 21 | 1938 | 137 | |
AK301768.1 | - | 19 - 20 | 1861 | 137 | |
BC011621.2 | - | 19 - 20 | 2093 | 137 |